missing translation for 'onlineSavingsMsg'
Learn More

β-Lapachone, Tocris Bioscience™

DNA topoisomerase I inhibitor

Brand:  Tocris (Bio-Techne) 129350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13439073

  • £401.31 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DNA topoisomerase I inhibitor. Exhibits a different inhibitory mechanism to camptothecin (Cat. No. 1100). Induces apoptosis in a variety of cell lines, including prostate cancer and promyelocytic leukemia cells; blocks the cell cycle in G0/G1. Also has effects on DNA topoisomerase II, NF-κB, AP-1, and JNK.
Specifications

Chemical Identifiers

4707-32-8
242.274
beta-lapachone, lapachone, beta, .beta.-lapachone, lapachone, 2,2-dimethyl-3,4-dihydro-2h-benzo h chromene-5,6-dione, unii-6n4fa2qq6a, 3,4-dihydro-2,2-dimethyl-2h-naphtho 1,2-b pyran-5,6-dione, 2h-naphtho 1,2-b pyran-5,6-dione, 3,4-dihydro-2,2-dimethyl, 6n4fa2qq6a
CHEBI:10429
CC1(CCC2=C(O1)C3=CC=CC=C3C(=O)C2=O)C
C15H14O3
QZPQTZZNNJUOLS-UHFFFAOYSA-N
3885
2,2-dimethyl-3,4-dihydrobenzo[h]chromene-5,6-dione

Specifications

50mg
4707-32-8
beta-lapachone, lapachone, beta, .beta.-lapachone, lapachone, 2,2-dimethyl-3,4-dihydro-2h-benzo h chromene-5,6-dione, unii-6n4fa2qq6a, 3,4-dihydro-2,2-dimethyl-2h-naphtho 1,2-b pyran-5,6-dione, 2h-naphtho 1,2-b pyran-5,6-dione, 3,4-dihydro-2,2-dimethyl, 6n4fa2qq6a
CC1(CCC2=C(O1)C3=CC=CC=C3C(=O)C2=O)C
242.274
CHEBI:10429
>98%
β-Lapachone
C15H14O3
QZPQTZZNNJUOLS-UHFFFAOYSA-N
2,2-dimethyl-3,4-dihydrobenzo[h]chromene-5,6-dione
3885
242.27
Videos
Documents