Learn More
LAP (Latency Associated peptide) Monoclonal Antibody (TW7-16B4), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£71.75 - £254.00
Specifications
Antigen | LAP (Latency Associated peptide) |
---|---|
Clone | TW7-16B4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15371400
|
Affymetrix eBioscience
46-9821-82 |
100 μg |
£254.00
100µg |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15361400
|
Affymetrix eBioscience
46-9821-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The TW7-16B4 monoclonal antibody reacts with mouse latency associated peptide (LAP, pro-TGF beta 1, LAP/TGF beta 1). Many different cells produce TGF beta and it mediates effects on the proliferation, differentiation and function of many cell types. TGF beta is synthesized as a precursor that contains LAP at the N-terminus and mature TGF beta at the C-terminus. Processing and cleavage of the precursor protein between amino acids 278 and 279 results in the formation of LAP dimers and TGF beta dimers that then non-covalently associate with each other to form the small latent TGF beta complex. LAP is secreted and can be found in the extracellular matrix. In addition, LAP can also be expressed on platelets and activated regulatory T cells. It is believed that this surface-expressed LAP is due to the binding of LAP to GARP (LRRC32), which is a transmembrane protein that is also found at high levels on platelets and activated regulatory T cells. Applications Reported: This TW7-16B4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TW7-16B4 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
LAP (Latency Associated peptide) Monoclonal antibody specifically detects LAP (Latency Associated peptide) in Mouse samples. It is validated for Flow Cytometry.Specifications
LAP (Latency Associated peptide) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P04202 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
TW7-16B4 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Mouse | |
21803 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.