Learn More
LAP (Latency Associated peptide) Monoclonal Antibody (VB3A9), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 16-9823-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: This VB3A9 monoclonal antibody recognizes human latency-associated peptide (LAP), also known as the TGF-beta propeptide. Human LAP is a homodimer of 65-75 kDa that is non-covalently associated with TGF-beta and this complex is also referred to as the small latent complex (SLC). LAP can be covalently associated with latent TGF-beta binding proteins (LTBP) through disulfide bonds, forming the large latent complex (LLC). The association of LAP with TGF-beta facilitates its secretion and also renders TGF-beta inactive. VB3A9 has been reported to cross-react with non-human primate LAP, but does not cross-react with mouse LAP. Applications Reported: This VB3A9 antibody has been reported for use in immunoblotting (WB) and immunoprecipitation. Applications Tested: This VB3A9 antibody has been tested by immunoblot analysis of human platelets under reducing conditions at 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
TGF beta-1 is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its prodomain. Activation of TGF-b1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF-beta, an aminoterminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP.Specifications
LAP (Latency Associated peptide) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P01137 | |
TGFB1 | |
Affinity chromatography | |
RUO | |
7040 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
VB3A9 | |
Functional Grade | |
TGFB1 | |
CED; DPD1; LAP; latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGFB; Tgfb1; Tgfb-1; TGFbeta; TGF-beta 1; TGFbeta1; TGF-beta1; TGF-beta-1; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor, beta 1; transforming growth factor-beta 1 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
For Research Use Only.