missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LanthaScreen™ TR-FRET PPAR delta Coactivator Assay Kit, goat
LanthaScreen™ TR-FRET PPAR delta Coactivator Assay Kit, goat
Brand: Invitrogen™ PV4685
Code : 50
Additional Details : Weight : 0.09000kg
Description
This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15134:The LanthaScreen™ TR-FRET PPAR (peroxisome proliferator activated receptor) delta Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential PPAR delta ligands as agonists of ligand-dependent coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human recombinant PPAR delta ligand-binding domain (PPAR delta-LBD) that is tagged with glutathione-S-transferase (GST), in a homogeneous mix-and-read assay format.
When running the LanthaScreen™ TR-FRET PPAR delta Coactivator Assay in agonist mode (to identify agonist compounds), PPAR delta-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-labeled coactivator peptide and Tb-labeled anti-GST antibody. After an incubation period at room temperature, the TR-FRET ratio of 520/495 is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the PPAR delta-coactivator peptide interaction, this ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (see Figure 1).
Contents and Storage:
The LanthaScreen™ TR-FRET PPAR delta Coactivator Assay Kit contains PPAR delta-LBD (GST) protein, fluorescently labeled C33 coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C or 4°C).
Specifications
Tb (Terbium) | |
Microplate Reader | |
Store in ultra-cold freezer (-68 to -85°C). | |
5467 | |
Dry Ice | |
LanthaScreen™ | |
PPAR delta |
Fluorescent | |
384-well Plate | |
Biochemical coregulator interaction | |
End Point | |
PPAR delta, NR1C2, PPARD | |
Co-Factor Interaction Assay, TR-FRET | |
TR-FRET PPAR Delta Coactivator Assay Kit |
For Research Use Only. Not for use in diagnostic procedures.