missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Eu-anti-His Tag Antibody Kit
LanthaScreen™ Eu-anti-His Tag Antibody Kit
£1010.00 - £13970.00
Specifications
Antigen | His |
---|---|
Concentration | 0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml |
Molecular Weight of Antigen | 150kDa |
Packaging | 1 Kit(s) |
Primary or Secondary | Primary |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10667044
|
Invitrogen
PV5596 |
25 μg |
£1010.00
Each |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105454
|
Invitrogen
PV5597 |
1 mg |
£13970.00
Each |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
LanthaScreen™ europium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.Specifications
His | |
150kDa | |
Primary | |
Liquid |
0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml | |
1 Kit(s) | |
Europium-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C. | |
RUO, RUO |
For Research Use Only. Not for use in diagnostic procedures.