Learn More
Thermo Scientific™ LanthaScreen™ Eu-anti-Biotin Antibody
LanthaScreen™ Eu-anti-Biotin Antibody
£852.00 - £12500.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10123078
|
Thermo Scientific™
PV5901 |
25μg |
£852.00
Each |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10214448
|
Thermo Scientific™
PV5900 |
1 mg |
£12500.00
Each |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A general reagent for use in assay development, the Europium-anti-biotin antibody binds to biotinylated peptide or physiological protein substrates.For Research Use Only. Not for use in diagnostic procedures.