missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum sputtering target, 76.2mm dia x 3.18mm thick, Thermo Scientific™

Catalog Number 15407135
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 7439-91-0
Molecular Formula La
MDL Number MFCD00011066
UN Number UN3208
Quantity 1 Ea.
Synonym in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
SMILES [La]
IUPAC Name lanthanum
Molecular Weight (g/mol) 138.91
PubChem CID 23926
ChEBI CHEBI:33336
Chemical Name or Material Lanthanum sputtering target, 76.2mm dia. x 3.18mm thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'