missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lanthanum Nickel eutectic pieces, REacton™, 99.9% (REO)

Brand:  Alfa Aesar™ 013399.18

Code : NEW

Additional Details : Weight : 0.05000kg Transport : UN number : 3178 Chem class : 4.1 Pack group : III

Product Code. 15486005

  • £70.60 / 50g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Specifications

La:Ni; 88:12 wt%
50g
Pieces
(REO)
UN3178
480°C
Lanthanum Nickel eutectic pieces
Reacton™
99.9%
MFCD00198177
Air & Moisture Sensitive
Videos
SDS