missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
39.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
Description
Lanthanum(III) nitrate is utilized as a starting material for the electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. It is used as a catalyst for the chemoselective preparation of cyclic and acyclic dithioacetals. It is used to prepare lanthanum aluminate (LAO) thin films, LaF3 nanocrystals, and bis(indolyl) methanes from indoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 40°C |
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 50 g |
Assay Percent Range | 99.99% (REO) |
Solubility Information | Soluble in water,alcohol and acids. |
IUPAC Name | lanthanum(3+) hexahydrate trinitrate |
Formula Weight | 433.01 (324.92 Anhydrous) |
Odor | Odorless |
Appearance | White crystalline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'