missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11306928
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
13709-38-1
F3La
195.90
MFCD00011073
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
lanthanum(3+) trifluoride
[F-].[F-].[F-].[La+3]
This item is not returnable. View return policy
13709-38-1
F3La
195.90
MFCD00011073
BYMUNNMMXKDFEZ-UHFFFAOYSA-K
lanthanum(3+) trifluoride
[F-].[F-].[F-].[La+3]

Lanthanum fluoride, anhydrous is employed extensively in the preparation and study of fluoride glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1493°C
Physical Form Lump
Quantity 250 g
Linear Formula LaF3
Solubility Information Insoluble in water. Soluble in strong mineral acids.
Formula Weight 195.9
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Sensitivity Hygroscopic
Density 5.936 g/mL
Assay (REO)
Chemical Name or Material Lanthanum(III) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'