Learn More
Lanthanum(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Description
Lanthanum(III) carbonate hydrate is used as a starting material for the preparation of lanthanum strontium manganite, which is used in solid oxide fuel cell applications. It is also involved in the preparation of high-temperature superconductors. It finds application in medicine as a phosphate binder. Further, it is used in the treatment of hyperphosphatemia. In addition to this, it is used for the tinting of glass, water treatment and as a catalyst for hydrocarbon cracking.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Lump/Powder |
Quantity | 50 g |
Merck Index | 14,5364 |
Solubility Information | Soluble in acids. Insoluble in water. |
Formula Weight | 457.85 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Density | 2.6 g/mL to 2.7 g/mL |
Assay | (REO) |
Show More |
RUO – Research Use Only