missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lanthanum foil, 0.62mm (0.024 in.) thick, 99.9% (REO)

Brand:  Alfa Aesar™ 000597.FF

Code : NEW

Additional Details : CAS Number : 7439-91-0 Weight : 0.01000kg Transport : UN number : 3208 Chem class : 1 Pack group : II


Quantity 25x25mm

 View more versions of this product

Product Code. 11356587

  • £100.59 / 25mm


Estimated Shipment date
from Supplier 28-02-2018
Add to basket


Specifications

Chemical Identifiers

Lanthanum foil
0.62mm (0.024 in.) thick
UN3208
7439-91-0
MFCD00011066

Specifications

Lanthanum foil
25x25mm
7439-91-0
99.9%
MFCD00011066
Mylar under Argon
≈2.4g/25x25mm
0.62mm (0.024 in.) thick
Foil
(REO)
UN3208
Silver-gray
Videos
SDS