missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Chloride, 7-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

£213.00

Chemical Identifiers

CAS 10099-58-8
Molecular Formula LaCl3·7H2O
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15587994 View Documents JT Baker
2255-05
100g Wide Mouth Amber Glass Bottle £213.00
100g View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    For Flame Enhancement in Atomic Absorption Spectroscopy
Specifications

Chemical Identifiers

10099-58-8
LaCl3·7H2O

Specifications

Lanthanum Chloride
10099-58-8
371.38
Wide Mouth Amber Glass Bottle
7-Hydrate
LaCl3·7H2O
BAKER ANALYZED™ ACS Reagent
100g