Alfa Aesar™ Lanthanum Aluminum Oxide Substrate, 10 x 10 x 1mm, Polished Two Sides, 100 Orientation, Alfa Aesar™


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 041435.KT

UNSPSC: 12141607

Code: NEW

Additional Details:
Additional Details: CAS Number: 12003-65-5 Weight: 0.01000kg


Quantity 1 ea.

Product Code. 11364148

Quantity Price
1 £ 122.64 / Each
Estimated Shipment date
from Supplier 09-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Lanthanum Aluminum Oxide Substrate
CAS 12003-65-5
Name Note Polished Two Sides
Molecular Formula LaAlO3
MDL Number MFCD00144550

Specification

Physical Form Plate
Color Yellow-brown
Quantity 1 ea.
MDL Number MFCD00144550
CAS 12003-65-5
Odor Odorless
Name Note Polished Two Sides
Molecular Formula LaAlO3
Chemical Name or Material Lanthanum Aluminum Oxide Substrate
Documentation

Technical documents