missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Lanthanum, AAS standard solution, Specpure™, La 1000μg/mL

Brand:  Alfa Aesar™ 088074.AP

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 500mL

 View more versions of this product

Product Code. 11306359

  • £64.10 / 500mL


Estimated Shipment date
from Supplier 20-03-2018
Add to basket

Specifications

Chemical Identifiers

Lanthanum, AAS standard solution
Specpure™
UN3264
La 1000μg/mL
MFCD00011066

Specifications

Lanthanum, AAS standard solution
Specpure™
La in 5% HNO3
UN3264
La 1000μg/mL
Liquid
MFCD00011066
500mL
Videos
SDS