missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanadelumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA542104
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Lanadelumab, also known as DX-2930, is a human IgG1 monoclonal antibody designed for subcutaneous self-injection. It is a fully human immunoglobulin, k-light-chain made in recombinant Chinese Hamster Ovary cells. The FDA and EU granted the designation of priority review, breakthrough therapy and orphan pharmaceutical for rare diseases based on the results of the reported clinical trials. Lanadelumab was developed by Shire and FDA approved on August 28, 2018. 5.Specifications
Lanadelumab | |
Recombinant Monoclonal | |
Unconjugated | |
DX-2930; X124-G01 | |
Human KLKB1 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
3.16 mg/mL | |
PBS with no preservative; pH 7.5 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |