Learn More
LAMTOR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552352
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VMFSSKERGR CTVINFVPLE APLRSTPRSR QVTEACGGEG RAVPLGSEPE WSVGGMEATL EQHLEDTMKN PSIVGVLCTD SQGLNLGCRG TLSDEHAGVI SVLAQQAAKL TSDPTDIPVV CLESDNGNIM IQKHD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
LAMTOR5 Polyclonal antibody specifically detects LAMTOR5 in Human samples. It is validated for Immunohistochemistry.Specifications
LAMTOR5 | |
Polyclonal | |
Unconjugated | |
LAMTOR5 | |
1110003H18Rik; HBV X-interacting protein; HBV X-interacting protein homolog; HBx-interacting protein; HBX-interacting protein homolog; HBXIP; hepatitis B virus x interacting protein; Hepatitis B virus X-interacting protein; hepatitis B virus x-interacting protein (9.6kD); hepatitis B virus X-interacting protein homolog; LAMTOR5; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 5; late endosomal/lysosomal adaptor, MAPK and MTOR activator 5; MGC71071; ragulator complex protein LAMTOR5; XIP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10542 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43504 | |
LAMTOR5 | |
Recombinant protein corresponding to Human LAMTOR5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only