missing translation for 'onlineSavingsMsg'
Learn More

LAMP5 Rat anti-Human, Alexa Fluor 488, Clone: 34.2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-9778-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15580297

  • £288.00 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The monoclonal antibody 34.2 recognizes human lysosome-associated membrane protein 5 (LAMP5), a member of the LAMP family. LAMP5 differs from other LAMP family members in that its expression is more restricted and is referred to as brain and dendritic cell-associated LAMP-like molecule (BAD-LAMP). In humans, BAD-LAMP is expressed in post-natal neurons and non-activated plasmacytoid dendritic cells. Expression in neurons is localized to intracellular vesicles distributed in specific microdomains along neurites and may play a role in endocytosis. BAD-LAMP is localized to the endoplasmic reticulum-Golgi intermediate compartment (ERGIC) of plasmacytoid dendritic cells and is lost upon TLR activation.
Specifications

Specifications

LAMP5
34.2
Alexa Fluor 488
Conjugated
IgG1, kappa
RUO
Primary
Human
Immunohistochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos
Documents

For Research Use Only.