Learn More
Lamotrigine, Tocris Bioscience™
Inhibits glutamate release. Anticonvulsant
Brand:  Tocris (Bio-Techne) 161110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13449793
  • £145.53 / 10mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Anticonvulsant. Inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents. Also blocks heterologously expressed and native α4β2 nAChRs with a similar affinity to Na+ channels.
Water-soluble Salt also available.
Specifications
Chemical Identifiers
84057-84-1
256.09
lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
CHEBI:6367
C1=CC(=C(C(=C1)Cl)Cl)C2=C(N=C(N=N2)N)N
C9H7Cl2N5
PYZRQGJRPPTADH-UHFFFAOYSA-N
3878
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
Specifications
10mg
84057-84-1
lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
C1=CC(=C(C(=C1)Cl)Cl)C2=C(N=C(N=N2)N)N
256.09
CHEBI:6367
>99%
Lamotrigine
C9H7Cl2N5
PYZRQGJRPPTADH-UHFFFAOYSA-N
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
3878
256.09