Learn More
Lamotrigine, Tocris Bioscience™
Inhibits glutamate release. Anticonvulsant
Brand:  Tocris (Bio-Techne) 161150
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13459793
  • £492.72 / 50mg
Estimated Shipment
02-10-2018
Add to basket
Description
Description
Anticonvulsant. Inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents. Also blocks heterologously expressed and native α4β2 nAChRs with a similar affinity to Na+ channels. Water-soluble salt available ( lamotrigine isethionate, Cat. No. 2289).
Specifications
Chemical Identifiers
84057-84-1
256.09
6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine
CHEBI:6367
C9H7Cl2N5
PYZRQGJRPPTADH-UHFFFAOYSA-N
3878
Specifications
Lamotrigine
>99%
6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine
C1=CC(=C(C(=C1)Cl)Cl)C2=C(N=C(N=N2)N)N
256.09
CHEBI:6367
50mg
84057-84-1
C9H7Cl2N5
PYZRQGJRPPTADH-UHFFFAOYSA-N
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
3878
256.09
Videos