missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine isethionate, Tocris Bioscience™

Inhibits glutamate release. Water-soluble salt of lamotrigine (Cat. No. 1611)

Brand:  Tocris (Bio-Techne) 228910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13460944

  • £133.65 / 10mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Water-soluble salt of lamotrigine (Cat. No. 1611). Displays anticonvulsant effects and inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents.
Specifications

Chemical Identifiers

113170-86-8
382.216
lamotrigine isethionate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine isethionate, 3,5-diamino-6-2,3-dichlorophenyl-1,2,4-triazine isethionate, acmc-1bsdt, isethionic acid; lamotrigine, lamotrigine isethionate hplc, 2-hydroxyethane-1-sulfonic acid-6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 1/1, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 2-hydroxyethanesulfonate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine mono 2-hydroxyethanesulfonate
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine;2-hydroxyethanesulfonic acid
C11H13Cl2N5O4S
CJIDZLNMKONKAD-UHFFFAOYSA-N
45039646
C1=CC(=C(C(=C1)Cl)Cl)C2=C(N=C(N=N2)N)N.C(CS(=O)(=O)O)O

Specifications

Lamotrigine isethionate
113170-86-8
lamotrigine isethionate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine isethionate, 3,5-diamino-6-2,3-dichlorophenyl-1,2,4-triazine isethionate, acmc-1bsdt, isethionic acid; lamotrigine, lamotrigine isethionate hplc, 2-hydroxyethane-1-sulfonic acid-6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 1/1, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 2-hydroxyethanesulfonate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine mono 2-hydroxyethanesulfonate
C1=CC(=C(C(=C1)Cl)Cl)C2=C(N=C(N=N2)N)N.C(CS(=O)(=O)O)O
382.216
382.22
10mg
C11H13Cl2N5O4S
CJIDZLNMKONKAD-UHFFFAOYSA-N
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine;2-hydroxyethanesulfonic acid
45039646
>99%
Videos