Learn More
Laminin beta-3 Monoclonal Antibody (CL3363), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA524655
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KLVTSMTKQL GDFWTRMEEL RHQARQQGAE AVQAQQLAEG ASEQALSAQE GFERIKQKYA ELKDRLGQSS MLGEQGARIQ SVKTEAEELF GETMEMMDRM KDMELELLRG SQAIMLRSAD LTGLEKRVEQ Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%. Binds to an epitope located within the peptide sequence MTKQLGDFWTRMEEL as determined by overlapping synthetic peptides.
Laminin beta-3 Monoclonal antibody specifically detects Laminin beta-3 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Laminin beta-3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13751 | |
LAMB3 | |
Recombinant protein corresponding to Human Laminin beta-3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL3363 | |
Unconjugated | |
LAMB3 | |
AI1A; BM600-125KDA; Epiligrin subunit bata; kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; LAM5; Lamb3; Laminin B1k chain; laminin beta 3; laminin chain; laminin S B3 chain; laminin subunit beta 3; laminin subunit beta-3; laminin, beta 3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); laminin-5 subunit beta; LAMNB1; Nicein subunit beta; nicein, 125kD; nicein, 125kDa; nicein-125k; nicein-125kDa; RP1-272L16.3 | |
Mouse | |
Protein A | |
RUO | |
3914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only