Learn More
Laminin alpha 4 Antibody (CL3185) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-42393AF750
Product Code. 30302613
  • £558.00 / 0.10mL
Description
Laminin alpha 4 Monoclonal antibody specifically detects Laminin alpha 4 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin alpha 4
Monoclonal
Alexa Fluor 750
DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha
This antibody was developed using a recombinant protein derived from Q16363, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL3185
50mM Sodium Borate
Mouse
Protein A purified
RUO
3910
Store at 4⪚C in the dark.
IgG1