missing translation for 'onlineSavingsMsg'
Learn More

Lactulose, 99+%, ACROS Organics™

Brand:  Acros Organics 225930250

Code : 29

Additional Details : CAS Number : 4618-18-2 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10378500

  • £96.00 / 25g


Estimated Shipment
18-05-2018
Add to basket

Specifications

Chemical Identifiers

Lactulose
99+%
342.28
≥99.0% (on anhydrous substance) (HPLC)
C12H22O11
4-O-β-D-Galactopyranosyl-α-D-fructose

Specifications

Lactulose
Crystalline Powder
99+%
342.28
4-O-β-D-Galactopyranosyl-α-D-fructose
-46.0° to -50.0° (20°C, 589nm) (c=5, H2O, ammonia) on anhydrous substance
− 47.00 (20.00°C c=5,H2O,12hr)
White to almost white
4618-18-2
≥99.0% (on anhydrous substance) (HPLC)
C12H22O11
Authentic
Glass bottle
3% max.
3.0 to 7.0 (6% aq. soln.)
25g
Videos
SDS