missing translation for 'onlineSavingsMsg'
Learn More

Lactose Monohydrate, Extra Pure, SLR, Fine Powder, Fisher Chemical™

Brand:  Fisher Chemical L/0200/65

Code : 21

Additional Details : CAS Number : 63-42-3 Weight : 5.00000kg

Packaging Plastic keg
Quantity 5kg

 View more versions of this product

Product Code. 10151430

  • £260.00 / 5kg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

63-42-3
71587

Specifications

Plastic keg
219°C
63-42-3
360.3g/mol
White
5kg
71587
Solid
Videos
SDS Documents
Certificates