missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, lithium salt, 99%, ACROS Organics™

Brand:  Acros Organics 413330050

Code : 29

Additional Details : CAS Number : 867-55-0 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10485651

  • £12.20 / 5g


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

Lactic acid, lithium salt
Extra Pure
99%
CH3CH(OH)CO2Li
MFCD00065399
867-55-0
≥98.5%
C3H5LiO3
96.01
Lactic acid lithium salt

Specifications

(10% aq. soln.), (10% aq. soln.) at 0nm, 0.07 max. at 260nm
(10% aq. soln.)
(10% aq. soln.) clear, colorless
Crystalline Powder
99%
CH3CH(OH)CO2Li
MFCD00065399
Lactic acid lithium salt
Glass bottle
White
Lactic acid, lithium salt
867-55-0
Extra Pure
≥98.5%
C3H5LiO3
96.01
03, IV, 633
Authentic
6 to 9 (1% soln.)
5g
Videos
SDS