missing translation for 'onlineSavingsMsg'
Learn More

Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals

Catalog Number p-7050295
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
50-21-5
C3H6O3
90.08
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(O)C(O)=O
This item is not returnable. View return policy
50-21-5
C3H6O3
90.08
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(O)C(O)=O

It is used most commonly by athletes to describe the intense pain felt during exhaustive exercise. Used to make cultured dairy products, as a food preservative, and to make chemicals. Useful for the creation of highly aligned electrospun poly-L-lactic acid fibers for nerve regeneration applications. Poly(lactic acid) nanocomposites with various organoclays shows useful application for gas permeability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 18°C
Boiling Point 122°C (15 mm)
Assay Percent Range 85.0 to 90.0%
Beilstein 1209341
Merck Index 14,5336
Solubility Information Soluble in water,ethanol,furfurol,alcohol and water. Partially soluble in diethyl ether. Practically insoluble in chloroform,petroleum ether,carbon disulfide.
Formula Weight 90.08
Grade ACS
Density 1.209 g/mL
Quantity 100 g
Flash Point >110°C (230°F)
Physical Form Liquid
Refractive Index 1.427
Chemical Name or Material Lactic acid, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'