Learn More
Lactic acid, ACS, 85.0-90.0% aq. soln., Thermo Scientific Chemicals
Description
It is used most commonly by athletes to describe the intense pain felt during exhaustive exercise. Used to make cultured dairy products, as a food preservative, and to make chemicals. Useful for the creation of highly aligned electrospun poly-L-lactic acid fibers for nerve regeneration applications. Poly(lactic acid) nanocomposites with various organoclays shows useful application for gas permeability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 18°C |
Boiling Point | 122°C (15 mm) |
Assay Percent Range | 85.0 to 90.0% |
Beilstein | 1209341 |
Merck Index | 14,5336 |
Solubility Information | Soluble in water,ethanol,furfurol,alcohol and water. Partially soluble in diethyl ether. Practically insoluble in chloroform,petroleum ether,carbon disulfide. |
Formula Weight | 90.08 |
Grade | ACS |
Density | 1.209 g/mL |
Quantity | 100 g |
Show More |
RUO – Research Use Only