missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 85%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5391489
232.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
50-21-5
C3H6O3
90.08
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(O)C(O)=O
This item is not returnable. View return policy
50-21-5
C3H6O3
90.08
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
612
CHEBI:78320
2-hydroxypropanoic acid
CC(O)C(O)=O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 90.08
Percent Purity 85%
Grade BAKER ANALYZED™ ACS Reagent
Quantity 500 mL
Flash Point 112.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.20kg
Chemical Name or Material Lactic Acid
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'