missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557627
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KEGKSNEKND FTKFKTEQEN EAKCRNSKPG KKKNDFEQGE LYLREKFENS IESLRLFKND PLFFKPGSQF LYSTFGYTLL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
LACTB Polyclonal antibody specifically detects LACTB in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
LACTB | |
Polyclonal | |
Unconjugated | |
LACTB | |
G24; Lact1; LACT-1; lactamase beta; lactamase, beta; Lactb; mitochondrial 39S ribosomal protein L56; mitochondrial ribosomal protein L56; MRPL56; serine beta lactamase-like protein; serine beta-lactamase-like protein LACTB, mitochondrial; UNQ843/PRO1781 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114294, 300803, 80907 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P83111, Q9EP89 | |
LACTB | |
Recombinant protein corresponding to Human LACTB | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only