missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, capsule filter, MPC insert, injection port

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Brand:  Thermo Scientific™ SS00160-I

Additional Details : Weight : 5.60000kg

 View more versions of this product

Product Code. 15959926

  • £1955.46 / 15 units
Estimated Shipment: 04-03-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL film. This BPC includes 3 ports (1 membrane capsule filter, MPC insert, pinch clamps, and an injection site). It is available in 1, 5, 10 and 20L capacities.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors

BPC port specifications:

  • Line 1: 0.2μm PES membrane capsule filter + pinch clamp. Tubing: C-Flex 374, length: 30.5cm (12 in.), ID x OD: 9.5 x 15.9mm (0.38 x 0.63 in.)
  • Line 2: MPC insert + cap, pinch clamp. Tubing: C-Flex 374, length: 91.

Specifications

Specifications

32 x 56.5 cm (12.6 x 22.3 in.)
Two 4-mil. poly bags
Labtainer™
15 per case
Irradiated
10 L
12.6 in.
Polyethylene
C-Flex
56.5 cm
ASI™ 26/77
Capsule filter and MPC insert + cap
Protective material around rough areas
Room Temperature
2.64 gal.
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
32 cm
BioProcess Container
22.3 in.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers