missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock connectors, injection site

Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.

Brand:  Thermo Scientific™ SS00009-I

Additional Details : Weight : 4.20000kg

 View more versions of this product

Product Code. 15969896

  • £1256.76 / 30 units
Estimated Shipment: 07-03-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions

Applications:

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquid
  • Collecting chromatography feed and fraction
  • Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications

Specifications

24.9 x 19.8 cm (9.8 x 7.8 in.)
One 4-mil. poly bag
Labtainer™
Room Temperature
0.264 gal.
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
24.9 cm
BioProcess Container
7.8 in.
ASI™ 28
Luer lock connectors, reducer and injection site
50 per case
Irradiated
1 L
9.8 in.
Ethyl Vinyl Acetate (EVA)
EVA
19.8 cm
Videos
SDS
Documents
Certificates
Special Offers

Special Offers