missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Laboratory Stainless Steel Crucible Lid

Complete a stainless steel crucible with a non-magnetic, bright finish lid. Nickel Electro™ Laboratory Stainless Steel Crucible Lid is flat with a depression for centering and includes an extension on one side that serves as a handle.

£57.70 - £71.65

Specifications

Material Stainless Steel
Quantity 10 Pk.
Autoclavable Yes
Product Type Specs Crucible lid
Shape Flat
View More Specs

Includes: DiameterMetricOuter

Products 4
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
11409609
View Documents Promotion Details
Nickel Electro™
165
40 mm
£60.55
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
 
11499599
View Documents Promotion Details
Nickel Electro™
164
35 mm
£57.70
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
 
11489599
View Documents Promotion Details
Nickel Electro™
163
30 mm
£67.30
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
 
11419609
View Documents Promotion Details
Nickel Electro™
167
50 mm
£71.65
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
 
Description

Description

  • Non-magnetic
  • Stamped with the word Stainless Steel to assist in the identification of material when in use
  • Flat shape with a depression for centering on crucibles
  • Includes an extension on one side that serves as a handle
  • Crevice free

Non magnetic, bright finish, crevice free lid, handle

Specifications

Specifications

Stainless Steel
Yes
Flat
10 Pk.
Crucible lid
Videos
SDS
Documents

Documents

Product Certifications