Learn More
Cytiva (formerly Pall Lab) Laboratory Manifold Replacement Valve
Replacement valve for microbiology manifold.
Brand:  Cytiva (formerly Pall Lab) 4894
Additional Details : Weight : 0.24700kg
Product Code. 17164861
  • £357.00 / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
Specifications
Specifications
Black Fluorocarbon Elastomer O-ring
Stainless Steel (316L)
Non-sterile