missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BMP51/BMP53 Label Maker Cartridge: B-427 Vinyl

Label Maker Cartridge with B-427 Vinyl labels for labeling facility, equipment, drawers and cabinets with warnings, instructions and reminders.

Brand:  Brady™ 131575

Code : NEW

Additional Details : Weight : 0.12470kg

Product Code. 15183329

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved
  • All-in-one cartridge includes label supply and R4300 Series ribbon
Specifications

Specifications

Permanent Acrylic
Non-sterile
0.004 in.
6.35 mm
Vinyl
BMP™51 Label Maker, BMP™53 Label Maker
1.1 x 0.25 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers