Labconco™ XPert™ Nano Enclosures: 6ft. Width, Extra Deep Without Built-in Ionizer

Keep hazardous powders and particulates contained during procedures

Overview
Brand: Labconco™

Manufacturer Part Number: 3888621

UNSPSC: 41110000

Code: 03

Additional Details:
Additional Details: Weight: 228.80000kg



Requires: Support base

Product Code. 11741529

Quantity Price
1 £ 15147.01 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Product Type Enclosure
Color Glacier White, Gray
Sash Opening (Metric) 30cm
Width (Metric) 183cm
Lighting Front Mounted Fluorescent Lamp
Length (Metric) Exterior 90.4cm
Volume (Metric) Exhaust 165.18 to 273.72lps
For Use With (Application) Nanoparticle manipulation and dry powder chemical handling
Dimensions (L x W x H) Exterior 35.6 x 72 x 47.5 in. (90.4 x 183 x 120.65cm)
Exhaust ULPA filter
Depth (Metric) Interior 76.2cm
Noise 61 to 63dBA
Height (Metric) Exterior 120.65cm
Material Steel
Voltage 230V
Width (Metric) Exterior 183cm
Width (Metric) Interior 179.83cm
Hertz 50Hz
Electrical Requirements 230V 50Hz
Depth (Metric) 90cm
Certifications/Compliance CE
Coating Epoxy Coated Aluminum