Labconco™ XPert™ Nano Enclosures: 6ft. Width, Extra Deep With Built-in Ionizer

Keep hazardous powders and particulates contained during procedures

Overview
Brand: Labconco™

Manufacturer Part Number: 3888662

UNSPSC: 41110000

Code: 03

Additional Details:
Additional Details: Weight: 232.00000kg



Requires: Support base

Product Code. 11751529

Quantity Price
1 £ 17346.61 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Product Type Enclosure
Voltage 230V
Certifications/Compliance CE
Coating Epoxy Coated Aluminum
Exhaust ULPA filter
Material Steel
Color Glacier White, Gray
Height (Metric) Exterior 120.65cm
Noise 61 to 63dBA
Width (Metric) Exterior 183cm
Sash Opening (Metric) 30cm
Electrical Requirements 230V 50Hz
Length (Metric) Exterior 90.4cm
Dimensions (L x W x H) Exterior 35.6 x 72 x 47.5 in. (90.4 x 183 x 120.65cm)
Hertz 50Hz
For Use With (Application) Nanoparticle manipulation and dry powder chemical handling
Width (Metric) Interior 179.83cm
Depth (Metric) Interior 76.2cm
Width (Metric) 183cm
Volume (Metric) Exhaust 165.18 to 273.72lps
Depth (Metric) 90cm
Lighting Front Mounted Fluorescent Lamp