Labconco™ XPert™ Nano Enclosures: 5ft. Width Without Built-in Ionizer

Keep hazardous powders and particulates contained during procedures

Overview
£9477.08 valid until 31/05/2017
Log in to get the promotional price



Brand: Labconco™

Manufacturer Part Number: 3887521

UNSPSC: 41110000

Code: 03

Additional Details:
Additional Details: Weight: 192.00000kg



Requires: Support base

Product Code. 11711519

Quantity Price
1 £ 11846.36 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Certifications/Compliance CE
Depth (Metric) Interior 59.18cm
Depth (Metric) 74cm
Dimensions (L x W x H) Exterior 29.1 x 60 x 47.5 in. (73.9 x 152 x 120.65cm)
Electrical Requirements 230V 50Hz
Exhaust ULPA filter
For Use With (Application) Nanoparticle manipulation and dry powder chemical handling
Height (Metric) Exterior 120.65cm
Hertz 50Hz
Length (Metric) Exterior 73.9cm
Noise 58 to 63dBA
Product Type Enclosure
Sash Opening (Metric) 30cm
Voltage 230V
Volume (Metric) Exhaust 136.86 to 226.53lps
Width (Metric) Exterior 152cm
Width (Metric) Interior 149.35cm
Width (Metric) 152.4cm
Coating Epoxy Coated Aluminum
Color Glacier White, Gray
Lighting Front Mounted Fluorescent Lamp
Material Steel
Special Offers