missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ XPert™ Nano Enclosures: 5ft. Width With Built-in Ionizer

Keep hazardous powders and particulates contained during procedures

Brand:  Labconco™ 3887562

UNSPSC : 41110000

Code : 03

Additional Details : Weight : 194.39999kg

 View more versions of this product

Product Code. 11721519

  • £14366.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Requires : Support base

Specifications

Specifications

CE
74cm
230V 50Hz
Nanoparticle manipulation and dry powder chemical handling
50Hz
58 to 63dBA
30cm
136.86 to 226.53lps
149.35cm
Epoxy Coated Aluminum
Front Mounted Fluorescent Lamp
59.18cm
29.1 x 60 x 47.5 in. (73.9 x 152 x 120.65cm)
ULPA filter
120.65cm
73.9cm
Enclosure
230V
152cm
152.4cm
Glacier White, Gray
Steel