Labconco™ XPert™ Nano Enclosures: 5ft. Width, Extra Deep Without Built-in Ionizer

Keep hazardous powders and particulates contained during procedures

Overview
Brand: Labconco™

Manufacturer Part Number: 3888521

Enclosure 1,520mm x 900mm x 970mm (w x d x h)XPert Nano, deep Labconco ULPA

UNSPSC: 41110000

Code: 03

Additional Details:
Additional Details: Weight: 204.80000kg



Requires: Support base

Product Code. 11721529

Quantity Price
1 £ 13627.56 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Product Type Enclosure
Certifications/Compliance CE
Depth (Metric) Interior 59.69cm
Depth (Metric) 74cm
Dimensions (L x W x H) Exterior 35.6 x 60 x 47.5 in. (90.4 x 152 x 120.65cm)
Electrical Requirements 230V 50Hz
Exhaust ULPA filter
For Use With (Application) Nanoparticle manipulation and dry powder chemical handling
Height (Metric) Exterior 120.65cm
Hertz 50Hz
Noise 58 to 63dBA
Sash Opening (Metric) 30cm
Voltage 230V
Volume (Metric) Exhaust 165.18 to 273.72lps
Width (Metric) Exterior 152cm
Width (Metric) Interior 179.83cm
Width (Metric) 152.4cm
Coating Epoxy Coated Aluminum
Color Glacier White, Gray
Lighting Front Mounted Fluorescent Lamp
Material Steel
Length (Metric) Exterior 90.4cm