Labconco™ XPert™ Nano Enclosures: 4ft. Width Without Built-in Ionizer

Keep hazardous powders and particulates contained during procedures

Overview
Brand: Labconco™

Manufacturer Part Number: 3887421

UNSPSC: 41110000

Code: 03

Additional Details:
Additional Details: Weight: 110.40000kg



Requires: Support base

Product Code. 11791509

Quantity Price
1 £ 7961.72 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket


Description and Specification

Specification

Product Type Enclosure
Hertz 50Hz
Width (Metric) Interior 118.87cm
Noise 58 to 61dBA
Depth (Metric) Interior 59.69cm
Certifications/Compliance CE
For Use With (Application) Nanoparticle manipulation and dry powder chemical handling
Lighting Front Mounted Fluorescent Lamp
Height (Metric) Exterior 97cm
Exhaust ULPA filter
Material Steel
Voltage 230V
Coating Epoxy Coated Aluminum
Width (Metric) 122cm
Dimensions (L x W x H) Exterior 29.1 x 48 x 38.3 in. (73.9 x 122 x 97cm)
Length (Metric) Exterior 73.9cm
Width (Metric) Exterior 122cm
Electrical Requirements 230V 50Hz
Color Glacier White, Gray
Depth (Metric) 74cm
Sash Opening (Metric) 24cm