missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ XPert™ Nano Enclosures: 2ft. Width Without Built-in Ionizer

Keep hazardous powders and particulates contained during procedures

Brand:  Labconco™ 3887221

Code : 03

Additional Details : Weight : 74.40000kg

 View more versions of this product

Product Code. 11751509

  • £5673.69 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Requires : Support base

Description

Description

Room air is pulled into the enclosure through the front, flows to the baffle and finally passes through a 99.99% ULPA exhaust filter before returning to the laboratory or cleanroom. Features built-in blower and Guardian™ airflow monitor. With built-in ionizer; separate filtered exhauster is not required.

  • Ergonomic air foil with aerodynamic Clean-Sweep™ airflow openings; side spring pin releases air foil for cleaning
  • Upper dilution air supply and containment sash foil with side-entry air foils
  • Zone-perforated, rear baffle pivots down and removes for cleaning
  • Exhaust ULPA filter, 99.99% efficient on 0.12μm particles; accessible from top for leak scanning of entire surface
  • True bag-in/bag-out filter disposal system (requires certified technician)
  • Built-in blower with dynamically balanced, vibration-isolated motorized impeller and speed control
  • 40% Blower reserve allows speed adjustment (by certified technician) as filter loads
  • Pressure gauge to monitor filter loading
  • Guardian Airflow Monitor continuously monitors airflow; features visual and audible alarms if airflow is outside setpoint range

Construction

  • Choice of interior depth (depending on width selected): 50cm (23.4 in.) or 76cm (30 in.)
  • Type 304 stainless-steel interior sides, baffle, air foil and integral work surface
  • Glacier white and gray, dry powder epoxy-coated aluminum and steel exterior
  • Prewired fluorescent lamp and switches for blower and light
  • 0.25 in. (0.6cm) Thick tempered safety glass front sash
  • 24cm (9.45 in.) Sash opening height on 2 ft. models
  • Ergonomically angled and hinged sash pivots upward, locks in up position; wiping seal contains contaminants
  • Two utility ports for pass-through of electrical cords
Specifications
  • Sash Opening: 9.4 in.
  • At 100fpm Exhaust: 145cfg
  • At 100fpm Noise: 53-58dbA
  • At 80fpm Exhaust: 115cfm
  • At 80fpm Noise: 50-56dbA
  • At 60fpm Exhaust: 80cfm
  • At 60fpm Noise: 48-53dbA

Ideal for nanoparticle manipulation and dry powder chemical handling

Specifications

Specifications

CE
74cm
230V 50Hz
Nanoparticle manipulation and dry powder chemical handling
50Hz
53 to 58dBA
24cm
61cm
61cm
Glacier White, Gray
Steel
59.69cm
29.1 x 24 x 38.3 in. (73.9 x 61 x 97cm)
ULPA filter
97cm
73.9cm
Enclosure
230V
57.91cm
Epoxy Coated Aluminum
Front Mounted Fluorescent Lamp
Videos
View accessory products