missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Washer Inserts

For use with Labconco FlaskScrubber™ and SteamScrubber™ Washers

Brand:  Labconco 4588600

Code : 03

Additional Details : Weight : 1.82000kg

 View more versions of this product

Product Code. 15179675

  • £248.01 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

240 x 10 to 12mm O.D. tubes
Each
Retainer top, Lower and Upper racks
Spindle, Standard
Culture Tube Insert
45.7cm
21.8cm
FlaskScrubber™, SteamScrubber™ glassware washer
Type 304 Stainless Steel
11.4cm
1.8kg
Videos