missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Telescoping Base Stands

For 4 ft. telescoping base stand

Brand:  Labconco™ 3811100

Code : 03

Additional Details : Weight : 4.55000kg

 View more versions of this product

Product Code. 15169695

  • £93.43 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Epoxy-coated 18 ga. steel
  • Hooks on lower rear horizontal cross member

Specifications

Specifications

25.14cm
Base stand
Hook on the lower rear horizontal cross member of the telescoping base stand
9.9D x 24 in. W
60.96cm
2 ft. telescoping base stand
Epoxy Coated Steel
Videos