missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Support Shelves: Telescoping Base Stand

For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations.

Brand:  Labconco™ 3811102

Code : 03

Additional Details : Weight : 6.36000kg

 View more versions of this product

Product Code. 15149695

  • £168.34 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations (pre-2014). Support Shelf provides 8.0 in. x 20.0 in. (20 x 51 cm) work surface to support remote equipment such as an aspirator pump within easy access of the biosafety cabinet. Powder-coated steel shelf mounts to left-side tubular frame of the base stand. Includes powder-coated steel shelf and fasteners. Not for use with SoLo Hydraulic Lift Base Stand. Installation is required.
Specifications

Specifications

25.14cm
Support Shelf
Provides work surface to support remote equipment
7kg
121.9cm
4 ft. telescoping base stand
Powder-coated 18 ga. steel