missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Labconco™ SteamScrubber™ Labware Inserts

Hold a variety of wider-mouth flasks, beakers and other labware in inverted position

Brand:  Labconco™ 4401501

Code : 03

Additional Details : Weight : 1.14000kg

 View more versions of this product

Product Code. 15169655

  • £141.84 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

22.35cm
Hold a variety of wider mouth flasks, beakers and other labware in inverted position
Type 304 Stainless Steel, Plastic
10.0
1.1kg
Each
FlaskScrubber™, SteamScrubber™ glassware washer
20.3cm
Labware Insert
49cm
Videos