Labconco™ Shelves for FreeZone™ Freeze-Dry Systems

Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications

Overview
Brand: Labconco™

Manufacturer Part Number: 7442100

UNSPSC: 41104704

Code: 50

Additional Details:
Additional Details: Weight: 0.50000kg



Product Code. 10613513

Quantity Price
1 £ 690.05 / Pack of 3
Estimated Shipment
20-04-2017
Add to basket


Description and Specification

Specification

Size Small
For Use With (Equipment) Freezone™ drying chambers
Diameter (Metric) 15.87cm
Weight (Metric) 0.7kg
Includes Three aluminum shelves with 0.25 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, and three rubber feet
For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
Height (Metric) 23.87cm
Documentation