Labconco™ Shelves for FreeZone™ Freeze-Dry Systems

Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications

Overview
Brand: Labconco™

Manufacturer Part Number: 7441700

UNSPSC: 41104704

Code: 50

Additional Details:
Additional Details: Weight: 0.50000kg



Product Code. 10113342

Quantity Price
1 £ 765.15 / Pack of 3
Estimated Shipment
22-06-2017
Add to basket


Description and Specification

Specification

Diameter (Metric) 25.4cm
For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
For Use With (Equipment) Freezone™ drying chambers
Height (Metric) 30.98cm
Includes Three aluminum shelves with 0.25 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, and three rubber feet
Size Large
Weight (Metric) 0.9kg
Documentation