missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Protector™ Standard Storage Cabinet
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
Brand: Labconco™ 9900600
Code : 03
Additional Details : Weight : 50.00000kg
Description
- Support loads up to 800 lb. (363kg)
- Durable epoxy-coated steel construction
- Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
- Manual-closing, non-locking door(s)
Specifications
White | |
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. | |
Epoxy-Coated Steel | |
Epoxy | |
60.96cm |
Single, Left Hinge, Manual Close | |
90.17 to 93.34cm | |
362.87kg | |
Standard Storage Cabinet | |
58.96kg |