missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Brand:  Labconco™ 9900300

UNSPSC : 41100000

Code : 03

Additional Details : Weight : 50.00000kg

 View more versions of this product

Product Code. 15109715

  • £1095.58 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

White
Cabinet, Sitting-Height
90.17 to 93.34cm
362.87kg
Standard Storage Cabinet
58.96kg
Single, Right Hinge, Manual Close
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
Epoxy-Coated Steel
Epoxy
60.96cm